Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Surgical Knife with Comfort Grip, Scalpel Handle

Catalog No. 22079780 Shop All Surgical Design Products
Change view
Click to view available options
Size:
# 3
# 4
Color:
Blue
Green
Orange
Pink
Purple
Red
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
22-079-780 # 4 Blue
22-079-785 # 4 Red
22-079-795 # 4 Purple
22-079-781 # 4 Pink
22-079-777 # 3 Pink
22-079-810 # 3 Orange
22-079-825 # 3 Green
22-079-822 # 3 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-079-780 Supplier Surgical Design™ Supplier No. 271BL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Surgical Design™ Surgical Knife Scalpel Handle used for sharp and precise dissection. The knife handle comes with enhanced PVC comfort and secure grip that increases precision during use.

  • Surgical Design™ Premium products exceed performance and quality expectations
  • Handles ergonomically designed for precision work
  • Handles offer color identification
  • Comfortable, reusable
  • Blades slide on smoothly and securely
  • Superb quality and workmanship
  • Premium surgical grade stainless steel
  • Corrosion protected by passivation and ultrasonic cleaning
  • Sterilizable by EtOH/Gamma Radiation
  • Fully autoclavable

Specifications

Product Type Scalpel Handle (High Precision)
For Use With (Application) 20 Series Scalpel Blades
Handle Type Comfort Grip
Size # 4
Disposable Reusable
Sterility Non-sterile
Certifications/Compliance ISO, GMP
Autoclavable Autoclavable
Color Blue
Graduated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.