Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Surgical Suture Kit
SCI_ED

Catalog No. S139052 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W):
8 x 7.8 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S139052 8 x 7.8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139052 Supplier Walter Products™ Supplier No. SK3D
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This complete, 21-piece tool kit has everything you need to practice mastering suturing techniques.

  • Resuable, 8 x 7.8 in. silicon suturing pad
  • Raised laceration types for practice that tolerate tension and pulling
  • Simulates anatomical structure of human tissue including 3 layers (skin, fat and muscle)
  • Acrylic base for easy handling and support
  • Threads of exceptionally smooth surface, superior tensile strength, and excellent knot security
  • Mixed variety of 75 cm medical grade surgical suture thread and curved needles: 6 Pk. Nylon, 6 Pk. Polyester, 6 Pk. Polypropylene, 6 Pk. Silk
  • Also includes: hegar needle holder, mosquito forceps, iris scissors, tooth forceps, scalpel and 10 scalpel blades

Specifications

Dimensions (L x W) 8 x 7.8 in.
For Use With (Application) Suturing Practice
Includes Practice Pad, Thread, Curved Needles, Forceps, Scissors, and Scalpel with Blades
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.