Learn More
Walter Products™ Surgical Suture Tool Kit
SCI_ED
Catalog No. S139051 Shop All Walter Products Products
Change view
Click to view available options
Suture Size (United States Pharmacopoeia):
4-0
Catalog No. Suture Size (United States Pharmacopoeia)
S139051 4-0
1 options
Catalog No. S139051 Supplier Walter Products™ Supplier No. SNK45
Add to Cart
Add to Cart
This quality, practice suturing kit includes all the tools you need to practice mastering suturing techniques.
  • Tool kit includes: 5 in. needle driver, 5 in. mosquito forcep, 4.5 in. iris scissors, 5 in. adson toothed forcep, scalpel handle (#3); 5 scalpel blades (#10) and leather case
  • 75 cm polypropylene, monofilament suture thread with superb strength that will not fatigue with normal flexing
  • Exceptionally smooth surface, superior tensile strength, and excellent knot security
  • Ultra sharp, 3/8 reverse cutting, stainless steel needle
  • Srong, perfectly opposed toothed jaws on needle driver for superior grip on the needle
  • Reliable ratchets that lock and unlock with ease
  • Comfortable finger loops ensure hours of pain-free practice
Specifications
For Use With (Application) Suturing Practice
Includes Needle Driver, Mosquito Forcep, Iris Scissors, Adson Toothed Forcep, Scalpel Handle, Scalpel Blades, Leather Case, Suture Thread with Needle
Thickness 18 mm
Suture Size (United States Pharmacopoeia) 4-0