Learn More
MediPurpose™ SurgiLance™ Lite Safety Lancet

Description
- Easy to use
- Minimizes pain
- Safe, single-use design
- 21 & 28 guage needles
- Eco-friendly design
- Easier for small hands
Specifications
Specifications
| Disposable | Single-use |
| Product Type | Fingerstick Lancet |
| Flow Type | Low (10 to 20μL) |
| Style | Lite |
| Depth (Metric) | 1.8mm |
| Color | Purple |
| For Use With (Application) | Glucose test |
| Needle Gauge | G21 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.