Learn More
MediPurpose™ SurgiLance™ Lite Safety Lancet

Description
- Easy to use
- Minimizes pain
- Safe, single-use design
- 21 & 28 guage needles
- Eco-friendly design
- Easier for small hands
Specifications
Specifications
| Disposable | Single-use |
| Product Type | Fingerstick Lancet |
| Flow Type | High (40 to 60μL) |
| Needle Gauge | G21 |
| Color | Pink |
| Depth (Metric) | 2.8mm |
| For Use With (Application) | Glucose test; Hematocrit tube |
| Style | Lite |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.