Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HR Pharmaceuticals Surgilube™ Surgical Lubricant

Catalog No. 22355175
Change view
Click to view available options
Packaging:
Laminated Tube with Flip-Top Cap
Metal Tube with Screw/Fez Cap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
22-355-175 Metal Tube with Screw/Fez Cap
22-355-176 Laminated Tube with Flip-Top Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-355-175 Supplier HR Pharmaceuticals Supplier No. FOU 0281020536
Add to Cart
Add to Cart
This item is not returnable. View return policy

This sterile, surgical lubricant is non-irritating to the skin, tissue, and mucous membranes. It is especially formulated for surgical and gynecological lubrication.

  • Surgilube™ sterile, surgical lubricant may be used where a sterile, water soluble, non-staining lubricating jelly is indicated.
  • Will not affect surgical instruments, rubber or plastics.

Specifications

Product Type Lubricant
Format Paste
For Use With (Equipment) Catheters, endoscopes, speculums, or other surgical instruments for body orifices
Packaging Metal Tube with Screw/Fez Cap
Weight (English) 4.25 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.