Learn More
World Precision Instrument Surgistain Rust and Stain Remover, 16 oz.
SDP
Supplier:  World Precision Instrument WP34572
Encompass
Removes rust, spots and stains from surgical instruments
For more then 20 years, hospitals worldwide have saved hundreds of thousands of dollars by using Surgistain to restore instruments. Used routinely as part of the Ruhof System, Surgistain will help maintain the life and efficiency of your instruments and reduce repair and replacements costs. If used on a monthly basis in conjunction with our enzymatic cleaners, Surgistain will restore the instruments' original finish and loosen box-locks and joints. It is an excellent choice for use on all ophthalmic and microsurgical instruments.
Catalog No. 50-253-8399
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.