Learn More
Surmodics Inc TMB One Component HRP Microwell Substrate, 1mL
SDP
Supplier:  Surmodics Inc TMBW010001
Encompass_Preferred
The BioFX™ one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution. Peroxidase reacts with the TMB substrate solution to produce a soluble blue reaction product that can be read at 370 nm or 620-650 nm for ELISA applications. Stop solutions can be added to prevent further color development and reading the absorbance values at the desired wavelengths.
Catalog No. NC0383033