Learn More
Terumo Medical Corporation SurTract™ Retractable Safety Syringe
Shop All Terumo Medical Corporation ProductsDescription
- Smooth and comfortable injection due to less plunger pressure
- Blunt needle allows change of needle for drawing up and prevents needle dullness
- Reduces medical waste due to lack of external plastic safety sheath
- Low dead space syringe reduces medication waste
- Customizable Needle Selection - SurTract™ safety needles are designed for use exclusively with SurTract™ safety syringes allowing you to create the combination you need
Specifications
Specifications
| Material | Polypropylene Ethylene Copolymer (Barrel), Polypropylene Ethylene Copolymer (Plunger) |
| Needle Gauge | No Needle Included |
| Latex Free | Yes |
| Product Type | Retractable Syringe without Needle |
| Disposable | Single-use |
| Volume (Metric) | 1 mL |
| Connection Type | Retractible |
| Graduated | Yes |
| Sterility | Sterile |