Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Surveyor™ Safety Eyewear

All day, pressure-free comfort

$72.20 - $232.00

Specifications

Certifications/Compliance ANSI Z87.1
Frame Color Black
Weight (Metric) 24 g
Lens Material Polycarbonate
Frame Material Polycarbonate
View More Specs

Products 4
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Coating Lens Tint Price Quantity & Availability  
19-155-773
View Documents
Pyramex™
SB6110ST
UV-treated Clear
Case of 12 for $72.20
Only null left
Add to Cart
 
19-155-774
View Documents
Pyramex™
SB6120S
Gray
Case of 12 for $162.50
Only null left
Add to Cart
 
19-155-775
View Documents
Pyramex™
SB6145S
Orange, Mirror
Case of 12 for $232.00
Only null left
Add to Cart
 
19-155-776
View Documents
Pyramex™
SB6170S
Silver, Mirror
Case of 12 for $196.70
Only null left
Add to Cart
 
Description

Description

  • Wrap-around, sunglass style fits all facial sizes
  • Cylinder lens demonstrates strength with style and protection
  • Lens provides unrestricted vision and protection against 99.9% harmful UV rays
  • Comfort-fit, rubber temples prevent eyewear from slipping to provide maximum performance
  • Specifications

    Specifications

    ANSI Z87.1
    24 g
    Polycarbonate
    Black
    Polycarbonate
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.