Learn More
ACROBiosystems ACROBiosystems Sus scrofa CD73 / NT5E Protein, His Tag
Supplier:  ACROBiosystems CD3S52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Sus scrofa CD73, His Tag (CD3-S52H3) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Ser 549 (Accession # XP_001927130.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.5 kDa.
Catalog No. 50-210-6816
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.