Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUSD2 Antibody (944812) [Janelia Fluor™ 525], R&D Systems™
SDP

Catalog No. NB647139
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB647139 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB647139 Supplier R&D Systems Supplier No. FAB9056JF525

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

SUSD2 Monoclonal antibody specifically detects SUSD2 in Human samples. It is validated for Immunohistochemistry,Western Blot

Specifications

Antigen SUSD2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 944812
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived human SUSD2, Met1-Ala785, Accession # Q9UGT4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 56241
Target Species Human
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.