Learn More
Mopec Suspended Cabinet, 3 Drawers
SDP
Catalog No. 21403146 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Drawer Cabinet
Catalog No. Product Type
21-403-146 Drawer Cabinet
1 options
Catalog No. 21-403-146 Supplier Mopec Supplier No. LC82215S
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • Each suspended cabinet furnished with complete hanging hardware
  • Includes (1) suspended cabinet with (3) drawers and hardware required to hang it
  • (2) drawers 4.9 in. H
  • (1) drawer 12.5 in. H
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • LC822-15S: 15.0 in. L x 22.0 in. W x 23.1 in. H
Specifications
Product Type Drawer Cabinet
Color Light Gray
Material Stainless Steel