Learn More
Med Vet International Ethicon Monocryl Suture, Size 2-0, FS-1, 36"", 12/Box
Supplier:  Med Vet International Y943G
Encompass_Preferred
The Ethicon Y943G suture is a high-quality absorbable Poliglecaprone 25 surgical suture. It measures in. in length, has a size of 2-0, and typically have an absorption rate of 90-120 days. This suture comes with a FS-1 reverse cutting needle and has 12 sutures in a box.
Catalog No. 50-209-2796
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.