Learn More
Med Vet International SUTPDS II PDO0CT-127
Supplier:  Med Vet International PDP340H
Encompass_Preferred
The Ethicon PDP340H suture is a high-quality absorbable Pds Plus Antibacterial Polydioxanone suture. It measures 27in in length, has a size of 0, and are dyed violet to provide greater visibility while suturing. This suture comes with a CT-1 36mm 1/2c Taper Needle and has 36 sutures in a box.
Catalog No. 50-255-9326
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.