Learn More
Med Vet International Oasis Gut Suture, Size 0, with NFSL Needle, 12/Box
Supplier:  Med Vet International MVN665
Encompass_Preferred
The Oasis Nylon suture with needle is a monofilament non-absorbable suture that is comparable to Ethicon™ Ethilon®. This Nylon suture has extremely high tensile strength that exceeds USP requirements. It provides excellent knot security, while the smooth surface allows for easy passage through the tissue. The thread color of this suture is blue and has a composition of polyamide. Size: 0. Needle Type: NFSL, 30mm 3/8 Circle Reverse Cutting. Length: 30in. Quantity: 12/box
Catalog No. 50-118-0618
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.