Learn More
Med Vet International Oasis PGA Suture Cassette, Size 1, Length of 15M, Each
Supplier:  Med Vet International PG01
Encompass_Preferred
The veterinary use Oasis PGA suture cassette is a braided synthetic absorbable suture. The PGA braided suture has a rapid absorption rate between 60 and 90 days. It retains 60 to 70% of its tensile strength at 14 days. This suture provides excellent pliability and holding strength. Size: 1. Length: 15M.
Catalog No. 50-264-3462
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.