Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 0, FSLX, 30in, Blue, 36/Box
Supplier:  Med Vet International 8690H
Encompass_Preferred
The Ethicon 8690H suture is a high-quality non-absorbable Prolene Polypropylene suture. It measures 30in in length, has a size of 0, and are dyed blue to provide greater visibility while suturing. This suture comes with a FSLX 36mm 3/8c Reverse Cutting Needle and has 36 sutures in a box.
Catalog No. 50-252-2494
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.