Learn More
Walter Products™ Suture Training Pad, 7.8 x 7.8 in.
SCI_ED
Catalog No. S139042 Shop All Walter Products Products
Change view
Click to view available options
Dimensions (L x W):
7.8 x 7.8 in.
Catalog No. Dimensions (L x W)
S139042 7.8 x 7.8 in.
1 options
Catalog No. S139042 Supplier Walter Products™ Supplier No. SP20013D
Add to Cart
Add to Cart
Reusable training pad for suturing technique.
  • Ideal for mastering suturing technique
  • Simulates anatomical structure of human tissue including 3 layers (skin, fat and muscle)
  • 12 pre-cut wounds of various widths and depths, as well as lots of space to add additional cuts
  • Strong vein forced to tolerate tension and pulling
  • Perfect for medical, nursing, and veterinary students
Specifications
Material Silicone
Dimensions (L x W) 7.8 x 7.8 in.
For Use With (Application) Suturing Practice
Includes Practice Pad and Manual