Learn More
Sklar Instruments Suture Wire Cutting Scissors, Sklar, 4-3/4 in., Blunt/Blunt, Angled, Serrated, Finger Rings, Stainless Steel. EA/1.
SDP
Catalog No. 501438014 Shop All Sklar Instruments Products
Encompass_Preferred
Suture Wire Cutting Scissors, Sklar, 4-3/4 in., Blunt/Blunt, Angled, Serrated, Finger Rings, Stainless Steel. Sklar's Suture Wire Cutting Scissors are non-ratcheted, finger ring scissors used for cutting sutures in preparation for removal. This product has straight shanks with angled, serrated jaws, blunt/blunt tips, and a length of 4-3/4 inches.
This item is not returnable. View return policy
Catalog No. 50-143-8014 Supplier Sklar Instruments Supplier No. 242342
Suture Wire Cutting Scissors, Sklar, 4-3/4 in., Blunt/Blunt, Angled, Serrated, Finger Rings, Stainless Steel. Sklar's Suture Wire Cutting Scissors are non-ratcheted, finger ring scissors used for cutting sutures in preparation for removal. This product has straight shanks with angled, serrated jaws, blunt/blunt tips, and a length of 4-3/4 inches.