Learn More
Med Vet International SUTUREMONOC-PLUS3-0SH27
Supplier:  Med Vet International MCP316H
Encompass_Preferred
The Ethicon MCP214H suture is a high-quality absorbable Monocryl Poliglecaprone 25 suture. It measures 27in in length, has a size of 3-0, and are dyed violet to provide greater visibility while suturing. This suture comes with a SH 26mm 1/2c Taper Needle and has 36 sutures in a box. Primary code substitute for discontinued code Y316H
Catalog No. 50-255-9557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.