Learn More
Med Vet International SUTUREMONOFYL0NCP-1VET USE
Supplier:  Med Vet International MVY967V
Encompass_Preferred
The Oasis veterinary use Monofyl suture with needle is a monofilament synthetic absorbable suture that is comparable to Ethicon™ Monocryl®. The Monofyl suture has an absorption rate between 91 and 119 days. Size: 0. Needle Type: NCP-1, 36mm 1/2 Circle Reverse Cutting Point. Length: 30in. Quantity: 12/box
Catalog No. 50-255-9355
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.