Learn More
Med Vet International Dynarex Braided (Pga) Sutures, Absorbable, Synthetic, Violet, 4-0, C3 Needle, 18", 12/Box
Supplier:  Med Vet International DYN9132
Encompass_Preferred
Used to hold body tissues together after an injury or surgery. Application generally involves using a needle with an attached length of thread. An absorbable, violet suture made from polyglycolic acid Braided for strength and easy handling and coated to allow for easy passage through tissue High tensile strength during healing period Available in various thread thickness and needle dimensions to accommodate a variety of procedures Sterile
Catalog No. 50-282-9612
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.