Learn More
Med Vet International SUTVIC PLGA 9102NO NDLE54
Supplier:  Med Vet International J618H
Encompass_Preferred
The Ethicon J618H suture is a high-quality absorbable Vicryl Polyglactin 910 suture. It measures 54in in length, has a size of 2, and are dyed violet to provide greater visibility while suturing. This suture comes with No Needle and has 36 sutures in a box.
Catalog No. 50-255-9571
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.