Learn More
Med Vet International Ethicon Coated Vicryl Plus Antibacterial Suture, Size 2-0, CT-1, 27"", 36/Box
Supplier:  Med Vet International VCP339H
Encompass_Preferred
The Ethicon VCP339H suture is a high-quality absorbable Polyglactin 910 surgical suture. It measures 27in. in length, has a size of 2-0, they are dyed violet to provide greater visibility while suturing, and typically have an absorption rate of 56-70 days. This suture comes with a CT-1 taper point needle and has 36 sutures in a box.
Catalog No. 50-209-2825
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.