Learn More
Suz12 Monoclonal Antibody (2AO9), Active Motif™

Description
Specifications
Specifications
| Antigen | Suz12 |
| Applications | ChIP Assay |
| Classification | Monoclonal |
| Clone | 2AO9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 |
| Gene | SUZ12 |
| Gene Accession No. | Q15022 |
| Gene Alias | 2610028O16Rik; AI195385; AU016842; AW536442; ChET 9 protein; CHET9; Chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; KIAA0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog (Drosophila); Suppressor of zeste 12 protein homolog; SUZ12; SUZ12 polycomb repressive complex 2 subunit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.