Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Suz12 Mouse anti-Mouse, Unconjugated, Clone: 4F7, MilliporeSigma™

Catalog No. MABE18761MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18761MI 100 μg
MABE18762MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18761MI Supplier MilliporeSigma Supplier No. MABE1876100UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suz12 Monoclonal antibody specifically detects Suz12 in Mouse samples. It is validated for Chromatin Immunoprecipitation (ChIP), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Suz12
Applications ChIP Assay, Western Blot
Classification Monoclonal
Clone 4F7
Conjugate Unconjugated
Gene Accession No. NP_954666.1
Gene Alias Polycomb protein Suz12;Suppressor of zeste 12 protein homolog
Gene Symbols Suz12, D11Ertd530e, Kiaa0160
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 191 amino acids from the C-terminal half of mouse Polycomb protein Suz12.
Molecular Weight of Antigen ∼85 kDa observed; 83.03 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone 4F7 is a mouse monoclonal antibody that detects Polycomb protein Suz12. It targets an epitope within the C-terminal region.
Target Species Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.