Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SV Internal Standard Mix, Restek™
SDP

Catalog No. 067073737 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Concentration:
2000 μg/mL
4000 μg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
06-707-3737 2000 μg/mL
06-707-3388 4000 μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-707-3737 Supplier Restek Corporation Supplier No. 31206
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Restek™ Semi-Volatile Mixtures - Mixes and Compounds
Concentration 2000 μg/mL
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.