Learn More
Genscript Corporation SV40 T-Ag-derived NLS peptide
Supplier:  Genscript Corporation RP20513
Encompass
This peptide, a nuclear localization signal DNA tagged to this peptide efficiently translocates into the cell nucleus.
Catalog No. NC2041772
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.