Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SVOP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553448 Supplier Invitrogen™ Supplier No. PA553448
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPIETKGRGL QESSHREWGQ EMVGRGMHGA GVTRSNSGSQ E Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.

SVOP gene ontology (GO) annotation includes transmembrane transporter activity and synaptic vesicle membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SVOP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SVOP
Gene Accession No. Q8N4V2
Gene Alias 1110030H18Rik; AI415691; msvop; SV2 related protein; SV2 related protein homolog; SV2-related protein; Svop; Synaptic vesicle 2-related protein
Gene Symbols SVOP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SVOP. Recombinant protein control fragment (Product #RP-91230).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55530
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.