Learn More
Med Vet International McKesson Hydrocolloid Dressing, 6 x 6 Inch, 1884, 10/Box
Supplier: Med Vet International 882983BX
Hydrocolloid Dressing McKesson 6 X 6 Inch Square Sterile The McKesson hydrocolloid dressing creates a moist environment that boosts healing It has a highly absorbent inner layer that absorbs wound exudates The thin hydrocolloid dressing is semi-transparent to provide visualization The outer layer is moisture resistant to prevent contamination It has a 7-day wear time 6 x 6 inches
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.