Learn More
Zymo Research Corporation Sample DNA/RNA Shield SafeCollectTM Swab Collection Kit, 2ml (1 Kit)
Supplier: Zymo Research Corporation R1161FS
Free Sample User-friendly collection for stabilizing the nucleic acid content of samples collected with a swab. DNA/RNA Shield completely inactivates harmful pathogens in the sample, and stabilizes the RNA in a sample for up to 30 days at room temperature. The SafeCollect design eliminates the risk of spilling the stabilization reagent during use, and is ideal for applications that require at-home self collection.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.