Learn More
3M™ Swab-Sampler For Pathogen Testing, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26765 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Media Type:
10 mL Buffered Peptone Water Broth
10 mL Letheen Broth
10 mL Neutralizing Buffer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
S26765 10 mL Letheen Broth
S26763 10 mL Buffered Peptone Water Broth
S26766 10 mL Neutralizing Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26765 Supplier 3M™ Supplier No. RS96010LET
Add to Cart
Add to Cart
Convenient and quick collection device for environmental sampling and testing over small surface areas
  • Ready to use for pathogen testing of environmental samples
  • Swab reaches into corners and hard-to-reach areas for increased accuracy
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Bottles have ample headspace for sample addition and vortexing
  • Self-contained swab and vial for increased productivity and efficiency
Specifications
For Use With (Application) Pathogen testing of environmental samples and testing over small surface areas
No. per Case 100/Cs.
Media Type 10 mL Letheen Broth
Product Type Swab