Learn More
3M™ Swab-Sampler, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26770 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Media Type:
1 mL letheen broth
4 mL letheen broth
4 mL neutralizing buffer
5 mL letheen broth
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
S26770 5 mL letheen broth
S26767 1 mL letheen broth
S26768 4 mL letheen broth
S26769 4 mL neutralizing buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S26770 Supplier 3M™ Supplier No. RS9605LET
Add to Cart
Add to Cart
Convenient and quick collection device for environmental sampling and testing over small surface areas
  • Letheen broth vial ready to use for pathogen testing of environmental samples
  • Swab reaches into corners and hard-to-reach areas for increased accuracy
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Bottles have ample headspace for sample addition and vortexing
  • Self-contained swab and vial for increased productivity and efficiency
Specifications
For Use With (Application) Environmental sampling and testing over small surface areas
No. per Case 100/Cs.
Media Type 5 mL letheen broth
Product Type Swab