Learn More
3M™ Swab-Sampler With D/E Neutralizing Broth, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26764 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
No. per Case:
100/Cs.
Catalog No. No. per Case
S26764 100/Cs.
1 options
Catalog No. S26764 Supplier 3M™ Supplier No. RS96010DE
Add to Cart
Add to Cart
Convenient and quick collection device for environmental sampling and testing over small surface areas
  • Ready to use for pathogen testing of environmental samples
  • Swab reaches into corners and hard-to-reach areas for increased accuracy
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Bottles have ample headspace for sample addition and vortexing
  • Self-contained swab and vial for increased productivity and efficiency
Specifications
Includes D/E neutralizing broth
For Use With (Application) Environmental testing and small surface areas
No. per Case 100/Cs.
Product Type Swab sampler