Learn More
MilliporeSigma™ Supelco™ Swagelok™ Back Ferrule, 404-1, Brass, 1/4 in.
Catalog No. 111005214 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
20/Pk.
Catalog No. Packaging Quantity
11-100-5214 20/Pk.
1 options
Catalog No. 11-100-5214 Supplier MilliporeSigma™ Supelco™ Supplier No. 22002
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.
The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Glass chromatographic column require brass back ferrule, PTFE front ferrule and brass nuts for adequate sealing.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 20/Pk.
Material Brass
Swagelok is a registered trademark of Swagelok Company