Learn More
MilliporeSigma™ Supelco™ Swagelok™ Fine Metering Valve Brass, angle arms 1/8 in.
Catalog No. 111010323 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Brass
Catalog No. Material
11-101-0323 Brass
1 options
Catalog No. 11-101-0323 Supplier MilliporeSigma™ Supelco™ Supplier No. 22114
Add to Cart
Edge
Add to Cart
Use for very accurate flow regulation. Install a flow measuring device downstream. Nuts and ferrules included. An optional vernier handle (Cat. No. 22122, order separately) is available for added convenience.
TRUSTED_SUSTAINABILITY
Specifications
Description Fine adjustment
Material Brass
Swagelok is a registered trademark of Swagelok Company