Learn More
MilliporeSigma™ Supelco™ Swagelok™ Fine Metering Valve Stainless Steel, straight arms 1/16 in.
Catalog No. 111008964 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
11-100-8964 Stainless Steel
1 options
Catalog No. 11-100-8964 Supplier MilliporeSigma™ Supelco™ Supplier No. 22121
Add to Cart
Add to Cart
Use for very accurate flow regulation. Install a flow measuring device downstream. Nuts and ferrules included. An optional vernier handle (Cat. No. 22122, order separately) is available for added convenience.
TRUSTED_SUSTAINABILITY
Specifications
Description Fine adjustment
Material Stainless Steel
Swagelok is a registered trademark of Swagelok Company