Learn More
MilliporeSigma™ Supelco™ Swagelok™ Nut, 202-1, Stainless Steel, 1/8 in.
Catalog No. 111011820 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
5/Pk.
Catalog No. Packaging Quantity
11-101-1820 5/Pk.
1 options
Catalog No. 11-101-1820 Supplier MilliporeSigma™ Supelco™ Supplier No. 22037
Add to Cart
Add to Cart
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.
The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Brass ferrules are used with copper tubing whereas stainless-steel ferrules are used with stainless-steel tubing.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 5/Pk.
Material Stainless Steel
Swagelok is a registered trademark of Swagelok Company