Learn More
MilliporeSigma™ Supelco™ Swagelok™ PTFE Ferrules PTFE back Ferrule, Swagelok™ for 1/16 in. Tubing
Catalog No. 111011784 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
5/Pk.
Catalog No. Packaging Quantity
11-101-1784 5/Pk.
1 options
Catalog No. 11-101-1784 Supplier MilliporeSigma™ Supelco™ Supplier No. 22069
Add to Cart
Add to Cart
Use PTFE ferrules instead of brass or stainless steel ferrules when attaching fittings to glass tubing, or to flexible tubing (such as PTFE or Tygon).
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 5/Pk.
Material PTFE
Swagelok is a registered trademark of Swagelok Company