Learn More
MilliporeSigma™ Supelco™ Swagelok™ PTFE Ferrules PTFE back Ferrule, Swagelok™ for 1/8 in. Tubing
Catalog No. 111011630 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Catalog No. Packaging Quantity
11-101-1630 10/Pk.
1 options
Catalog No. 11-101-1630 Supplier MilliporeSigma™ Supelco™ Supplier No. 22059
Add to Cart
Add to Cart
Use PTFE ferrules instead of brass or stainless steel ferrules when attaching fittings to glass tubing, or to flexible tubing (such as PTFE or Tygon).
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 10/Pk.
Material PTFE
Swagelok is a registered trademark of Swagelok Company