Learn More
MilliporeSigma™ Supelco™ Swagelok™ Reducing Union, 400-6-2, Stainless Steel, 1/4 in., 1/8 in.
Catalog No. 111005131 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-5131 1/Pk.
1 options
Catalog No. 11-100-5131 Supplier MilliporeSigma™ Supelco™ Supplier No. 22031
Add to Cart
Add to Cart
Reducing unions of Swagelok™™ type are mostly suitable as metal column terminators.
All original reducing unions have a bore in the centre of about 6 mm in length and an internal diameter of 1.2 mm. Moreover, reducing unions have a large bore at the non-reducing site. Modification is necessary in reducing union in order to minimize peak broadening, pressure resistance and to ease the installation process.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Material Stainless Steel
Swagelok is a registered trademark of Swagelok Company