Learn More
MilliporeSigma™ Supelco™ Swagelok™ Union Cross, 400-4, Brass, 1/4 in.
Catalog No. 111005104 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
11-100-5104 1/Pk.
1 options
Catalog No. 11-100-5104 Supplier MilliporeSigma™ Supelco™ Supplier No. 22686
Add to Cart
Add to Cart
Union cross generally divides the eluent flow into three directions which is coming from the pump. In gas system, pure gases from mass flow controllers are converged at the union cross and in turn flow into different size tubing to generate turbulence which produce homogenized gas.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Material Brass
Swagelok is a registered trademark of Swagelok Company