Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Swan™ Hydrogen Peroxide Solution

Catalog No. 19090693 Shop All Medique Products
Change view
Click to view available options
Volume (English):
8 oz.
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-090-693 16 oz.
19-090-694 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-090-693 Supplier Medique Supplier No. 25711
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For treatment of minor cuts and abrasions.

  • Topical local antiseptic

Specifications

For Use With (Application) Topical antiseptic treatment of minor cuts and abrasions
Product Type Hydrogen Peroxide Solution
Format Bottle
Quantity 1 Ea.
Volume (English) 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.