Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System
Catalog No. 60180792 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only
Includes:
SWAP-IT Interface only
Catalog No. For Use With (Equipment) Includes
60-180-792 DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only SWAP-IT Interface only
1 options
Catalog No. 60-180-792 Supplier Thermo Scientific™ Supplier No. 60180792
Make column changeovers quick and easy. Use this GC-MS interface system to switch capillary columns without the need to vent your mass spectrometer (MS).
Make column changeovers quick and easy. The Thermo Scientific™ SWAP-IT™ GC-MS Interface system is a powerful tool that enables capillary column changes without the need to vent your mass spectrometer (MS). It uses a specially deactivated, robust stainless steel transfer capillary as an interface between the analytical column and the MS detector. The small inner diameter of transfer capillary ensures minimum exposure to air and water when the analytical column is disconnected. A helium purge is not required. Finger-tight connections make it quick and easy without the need for tools.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type GC-MS Interface Unit
Description SWAP-IT GC-MS Interface Unit
Includes SWAP-IT Interface only
For Use With (Equipment) DSQ GC-MS, DSQ II GC-MS, PolarisQ GC-MS, ITQ GC-MS Only