Learn More
Kingfisher Biotech Inc Swine Annexin V Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1796S005
Encompass
The Swine Annexin V yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine Annexin V applications are for cell culture, ELISA standard, and Western Blot Control. Swine Annexin V yeast-derived recombinant protein can be purchased in multiple sizes. Swine Annexin V Specifications: (Molecular Weight: 36.0 kDa) (Amino Acid Sequence: MAQVLRGTVT DFPGFDERAD AETLRKAMKG LGTDEESILT LLTSRSNAQR QEIAVAFKTL FGRDLLDDLK SELTGKFEKL IVALMKPSRL YDAYELKHAL KGAGTDEKIL TEIIASRTPE ELRAIKQVYE EEYGSSLEDD VVGDTSGYYQ RMLVVLLQAN RDPDGRIDEA QVEQDAQALF QAGELKWGTD EEKFITILGT RSVSHLRRVF DKYMTISGFQ IEETIDRETS GNLEQLLLAV VKSIRSIPAY LAETLYYAMK GAGTDDHTLI RVMVSRSEID LFNIRKEFRK NFATSLYSMI KGDTSGDYKK ALLLLCGGED D (321)) (Gene ID: 100521982). For research use only. Made in the USA
Catalog No. 50-253-2762
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.