Learn More
Kingfisher Biotech Inc Swine/Bat Pleiotrophin Recombinant Protein
Supplier: Kingfisher Biotech Inc RP2001S100
The Swine/Bat Pleiotrophin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine/Bat Pleiotrophin applications are for cell culture, ELISA standard, and Western Blot Control. Swine/Bat Pleiotrophin yeast-derived recombinant protein can be purchased in multiple sizes. Swine/Bat Pleiotrophin Specifications: (Molecular Weight: 15.3 kDa) (Amino Acid Sequence: GKKEKPEKKV KKSDCGEWQW SVCVPTSGDC GLGTREGTRT GAECKQTMKT QRCKIPCNWK KQFGAECKYQ FQAWGECDLN TALKTRTGSL KRALHNADCQ KTVTISKPCG KVTKPKPQAE SKKKKKEGKK QEKMLD (136)) (Gene ID: 397609 (swine)). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.