Learn More
Kingfisher Biotech Inc Swine CCL17 Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2306S005
Encompass
The Swine CCL17 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine CCL17 applications are for cell culture, ELISA Standard, ELISpot Control, and Western Blot Control. Swine CCL17 yeast-derived recombinant protein can be purchased in multiple sizes. Swine CCL17 Specifications: (Molecular Weight:9.0 kDa) (Amino Acid Sequence:ARGTNVGREC CLEYFKGAIP LRKLVGWYQT SDECTRDAIV LVTFQGRAIC SDPKDTWVKK AIRYLKKNMK PQDPVTQKS). For research use only. Made in the USA.
Catalog No. 50-296-4659
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.