Learn More
Kingfisher Biotech Inc Swine CD40 Ligand (CD40LG) Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP0936S100
Encompass_Preferred
The Swine CD40 Ligand yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine CD40 Ligand applications are for cell culture, ELISA standard, and Western Blot Control. The Swine CD40 Ligand yeast-derived recombinant protein can be purchased in multiple sizes. Swine CD40 Ligand Specifications: (Molecular Weight: 16.0 kDa) (Amino Acid Sequence: MHKGDQDPQI AAHVISEASS KTASVLQWAP KGYYTLSTNL VTLENGRQLA VKRQGIYYIY AQVTFCSNRD AAGQAPFIAS LCLRSPSGSE RILLRAANTH SSSKPCGQQS IHLGGVFELQ PGASVFVNVT DPSQVSHGTG FTSFGLLKL (149)) (Gene ID: 397231). For research use only. Made in the USA
Catalog No. 50-241-3992
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.