Learn More
Kingfisher Biotech Inc Swine IFN alpha 1 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP0010S025
Encompass_Preferred
The Swine IFN alpha 1 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Swine IFN alpha 1 applications are for cell culture, ELISA standard, and Western Blot Control. The Swine IFN alpha 1 yeast-derived recombinant protein can be purchased in multiple sizes. Swine IFN alpha 1 Specifications: (Molecular Weight: 19.0 kDa) (Amino Acid Sequence: CDLPQTHSLA HTRALRLLAQ MRRISPFSCL DHRRDFGSPH EAFGGNQVQK AQAMALVHEM LQQTFQLFST EGSAAAWNES LLHQFCTGLD QQLRDLEACV MQGAGLEGTP LLEEDSILAV RKYFHRLTLY LQEKSYSPCA WEIVRAEVMR SFSSSRNLQD RLRKKE (166)) (Gene ID: 397686). For research use only. Made in the USA
Catalog No. 50-241-3394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.